Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
Chromosome Location
Chr.6: 49430360 - 49463328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005249143.3 XP_005249200.1 1 - 2 128 65
GenBank mRNA BX647789.1 - 1 - 2 48 65

View Full Product Details