Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC3
Entrez Gene ID
116138
Gene Name
kelch domain containing 3
Gene Aliases
PEAS, dJ20C7.3
Chromosome Location
Chr.6: 43014103 - 43021298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057161.3 NP_476502.1 6 - 7 1037 95
NR_040101.1 5 - 6 921 95
GenBank mRNA AB055925.1 - 6 - 7 901 95
AK097719.1 - 4 - 5 763 95
AK290384.1 - 6 - 7 893 95
AK302336.1 - 5 - 6 676 95
AK308917.1 - 6 - 7 923 95
BC000295.2 - 6 - 7 857 95
BC001789.1 - 6 - 7 872 95
BC001793.2 - 6 - 7 835 95
BC007296.1 - 6 - 7 887 95
BC009460.1 - 6 - 7 885 95
BC021546.1 - 6 - 7 874 95
BC041793.1 - 5 - 6 775 95
BC045612.1 - 5 - 6 775 95

View Full Product Details