The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000236.2 | NP_000227.2 | 4 - 5 | 630 | 69 |
XM_005254372.1 | XP_005254429.1 | 5 - 6 | 678 | 69 | |
XM_005254374.4 | XP_005254431.2 | 4 - 5 | 1972 | 69 | |
XM_006720502.3 | XP_006720565.1 | 4 - 5 | 575 | 69 | |
XM_017022176.1 | XP_016877665.1 | 4 - 5 | 1987 | 69 | |
GenBank mRNA | AK290062.1 | - | 6 - 7 | 1181 | 69 |
AK292631.1 | - | 6 - 7 | 1184 | 69 | |
AK293324.1 | - | 3 - 4 | 432 | 69 | |
AK315306.1 | - | 4 - 5 | 809 | 69 | |
AV645541.1 | - | 1 - 2 | 60 | 69 | |
BC132825.1 | - | 4 - 5 | 608 | 69 | |
BC136495.1 | - | 4 - 5 | 608 | 69 | |
BC146659.1 | - | 5 - 6 | 853 | 69 | |
D83548.1 | - | 4 - 5 | 614 | 69 | |
J03540.1 | - | 4 - 5 | 577 | 69 | |
J03895.1 | - | 4 - 5 | 580 | 69 | |
X07228.1 | - | 4 - 5 | 630 | 69 |