The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000236.2 | NP_000227.2 | 5 - 6 | 864 | 144 |
XM_005254372.1 | XP_005254429.1 | 6 - 7 | 912 | 144 | |
XM_005254374.4 | XP_005254431.2 | 5 - 6 | 2206 | 144 | |
XM_006720502.3 | XP_006720565.1 | 5 - 6 | 809 | 144 | |
XM_017022176.1 | XP_016877665.1 | 5 - 6 | 2221 | 144 | |
GenBank mRNA | AK290062.1 | - | 7 - 8 | 1415 | 144 |
AK292631.1 | - | 7 - 8 | 1418 | 144 | |
AK293324.1 | - | 4 - 5 | 666 | 144 | |
AK309448.1 | - | 4 - 5 | 664 | 144 | |
AK315306.1 | - | 5 - 6 | 1043 | 144 | |
AV645541.1 | - | 2 - 3 | 294 | 144 | |
BC132825.1 | - | 5 - 6 | 842 | 144 | |
BC136495.1 | - | 5 - 6 | 842 | 144 | |
BC146659.1 | - | 6 - 7 | 1087 | 144 | |
D83548.1 | - | 5 - 6 | 848 | 144 | |
J03540.1 | - | 5 - 6 | 811 | 144 | |
X07228.1 | - | 5 - 6 | 864 | 144 |