The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138295.3 | NP_612152.1 | 42 - 43 | 6510 | 124 |
XM_017011798.1 | XP_016867287.1 | 43 - 44 | 12915 | 124 | |
GenBank mRNA | AB061683.1 | - | 42 - 43 | 6476 | 124 |