The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004792.2 | NP_004783.2 | 8 - 9 | 628 | 144 |
XM_005246966.1 | XP_005247023.1 | 8 - 9 | 585 | 144 | |
XM_005246967.1 | XP_005247024.1 | 8 - 9 | 632 | 144 | |
XM_017005302.1 | XP_016860791.1 | 6 - 7 | 424 | 144 | |
GenBank mRNA | AK307552.1 | - | 5 - 6 | 419 | 144 |
BC001555.1 | - | 9 - 10 | 588 | 144 | |
BC063711.1 | - | 8 - 9 | 730 | 144 | |
BC073875.1 | - | 7 - 8 | 483 | 144 | |
BC109373.1 | - | 8 - 9 | 632 | 144 | |
BC111693.1 | - | 7 - 8 | 570 | 144 | |
U40763.1 | - | 9 - 10 | 893 | 144 | |
X99717.1 | - | 8 - 9 | 565 | 144 |