The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271534.1 | NP_001258463.1 | 16 - 17 | 3472 | 79 |
NM_001389.3 | NP_001380.2 | 16 - 17 | 3472 | 79 | |
NR_073202.1 | 16 - 17 | 3472 | 79 | ||
XM_017028281.1 | XP_016883770.1 | 13 - 14 | 2527 | 79 | |
GenBank mRNA | AB209136.1 | - | 16 - 17 | 3055 | 79 |
AF023449.1 | - | 15 - 16 | 2978 | 79 | |
AF023450.1 | - | 16 - 17 | 3472 | 79 | |
AF217525.1 | - | 16 - 17 | 3498 | 79 | |
AF289030.1 | - | 12 - 13 | 2276 | 79 |