The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271534.1 | NP_001258463.1 | 28 - 29 | 5373 | 75 |
NM_001389.3 | NP_001380.2 | 28 - 29 | 5373 | 75 | |
NR_073202.1 | 28 - 29 | 5182 | 75 | ||
XM_017028281.1 | XP_016883770.1 | 25 - 26 | 4428 | 75 | |
GenBank mRNA | AB209136.1 | - | 28 - 29 | 4956 | 75 |
AF023449.1 | - | 27 - 28 | 4879 | 75 | |
AF023450.1 | - | 28 - 29 | 5182 | 75 | |
AF217525.1 | - | 28 - 29 | 5399 | 75 | |
AF289030.1 | - | 24 - 25 | 4177 | 75 |