The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271534.1 | NP_001258463.1 | 12 - 13 | 3008 | 77 |
NM_001389.3 | NP_001380.2 | 12 - 13 | 3008 | 77 | |
NR_073202.1 | 12 - 13 | 3008 | 77 | ||
XM_017028281.1 | XP_016883770.1 | 9 - 10 | 2063 | 77 | |
GenBank mRNA | AB209136.1 | - | 12 - 13 | 2591 | 77 |
AF023449.1 | - | 11 - 12 | 2514 | 77 | |
AF023450.1 | - | 12 - 13 | 3008 | 77 | |
AF217525.1 | - | 12 - 13 | 3034 | 77 | |
AF289030.1 | - | 8 - 9 | 1812 | 77 |