The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013369.3 | NP_037501.2 | 1 - 2 | 479 | 107 |
NM_175867.2 | NP_787063.1 | 1 - 2 | 479 | 107 | |
GenBank mRNA | AF194032.1 | - | 1 - 2 | 479 | 107 |
BC002560.2 | - | 1 - 2 | 153 | 107 |