Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLCO1A2
Entrez Gene ID
6579
Gene Name
solute carrier organic anion transporter family member 1A2
Gene Aliases
OATP, OATP-A, OATP1A2, SLC21A3
Chromosome Location
Chr.12: 21264600 - 21403657 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011520820.2 XP_011519122.1 2 - 3 1680 124
GenBank mRNA AF085224.1 - 2 - 3 272 124

View Full Product Details