Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HDAC9
- Entrez Gene ID
-
9734
- Gene Name
- histone deacetylase 9
- Gene Aliases
-
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
- Chromosome Location
-
Chr.7: 18086942 - 19003745 on Build GRCh38
View Full Product Details