Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDAC9
Entrez Gene ID
9734
Gene Name
histone deacetylase 9
Gene Aliases
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location
Chr.7: 18086942 - 19003745 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204144.2 NP_001191073.1 7 - 8 1027 111
NM_001204145.2 NP_001191074.1 5 - 6 719 111
NM_001204146.2 NP_001191075.1 5 - 6 897 111
NM_001204148.2 NP_001191077.1 4 - 5 694 111
NM_001321868.1 NP_001308797.1 6 - 7 967 111
NM_001321869.1 NP_001308798.1 6 - 7 976 111
NM_001321870.1 NP_001308799.1 6 - 7 967 111
NM_001321871.1 NP_001308800.1 6 - 7 982 111
NM_001321872.1 NP_001308801.1 6 - 7 976 111
NM_001321874.1 NP_001308803.1 5 - 6 897 111
NM_001321875.1 NP_001308804.1 5 - 6 903 111
NM_001321877.1 NP_001308806.1 5 - 6 897 111
NM_001321878.1 NP_001308807.1 5 - 6 888 111
NM_001321885.1 NP_001308814.1 5 - 6 912 111
NM_001321886.1 NP_001308815.1 6 - 7 947 111
NM_001321891.1 NP_001308820.1 5 - 6 888 111
NM_001321893.1 NP_001308822.1 6 - 7 785 111
NM_001321894.1 NP_001308823.1 6 - 7 794 111
NM_001321895.1 NP_001308824.1 5 - 6 728 111
NM_001321896.1 NP_001308825.1 6 - 7 1066 111
NM_001321897.1 NP_001308826.1 6 - 7 794 111
NM_001321898.1 NP_001308827.1 5 - 6 743 111
NM_001321899.1 NP_001308828.1 6 - 7 794 111
NM_001321900.1 NP_001308829.1 5 - 6 888 111
NM_001321902.1 NP_001308831.1 5 - 6 728 111
NM_014707.3 NP_055522.1 5 - 6 719 111
NM_058176.2 NP_478056.1 4 - 5 574 111
NM_178423.2 NP_848510.1 5 - 6 719 111
NM_178425.3 NP_848512.1 4 - 5 583 111
XM_011515629.2 XP_011513931.1 5 - 6 677 111
XM_011515631.2 XP_011513933.1 6 - 7 991 111
XM_011515634.2 XP_011513936.1 5 - 6 677 111
XM_011515635.2 XP_011513937.1 6 - 7 1543 111
XM_011515636.2 XP_011513938.1 5 - 6 1264 111
XM_011515637.2 XP_011513939.1 5 - 6 688 111
XM_011515638.2 XP_011513940.1 5 - 6 1258 111
XM_011515639.2 XP_011513941.1 5 - 6 1455 111
XM_011515641.2 XP_011513943.1 6 - 7 1540 111
XM_011515642.2 XP_011513944.1 4 - 5 2269 111
XM_011515644.2 XP_011513946.1 6 - 7 1515 111
XM_011515645.2 XP_011513947.1 4 - 5 2254 111
XM_011515646.2 XP_011513948.1 4 - 5 2245 111
XM_011515648.2 XP_011513950.1 5 - 6 1273 111
XM_011515649.2 XP_011513951.1 5 - 6 1477 111
XM_017012826.1 XP_016868315.1 5 - 6 1249 111
XM_017012827.1 XP_016868316.1 6 - 7 1332 111
XM_017012828.1 XP_016868317.1 5 - 6 1457 111
GenBank mRNA AB018287.1 - 5 - 6 683 111
AF124924.1 - 2 - 3 241 111
AJ459808.1 - 5 - 6 683 111
AK097178.1 - 5 - 6 677 111
AK296214.1 - 5 - 6 629 111
AK297404.1 - 5 - 6 673 111
AK304298.1 - 7 - 8 1020 111
AK304343.1 - 4 - 5 614 111
AK316109.1 - 5 - 6 848 111
AY032737.1 - 4 - 5 533 111
AY032738.1 - 4 - 5 533 111
AY197371.1 - 4 - 5 542 111
BC111735.1 - 4 - 5 541 111
BC150328.1 - 5 - 6 694 111
BC152405.1 - 5 - 6 683 111
CR749550.1 - 4 - 5 650 111

View Full Product Details