Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDAC9
Entrez Gene ID
9734
Gene Name
histone deacetylase 9
Gene Aliases
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location
Chr.7: 18086942 - 19003745 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321868.1 NP_001308797.1 22 - 23 3096 86
NM_001321877.1 NP_001308806.1 21 - 22 3026 86
NM_001321897.1 NP_001308826.1 22 - 23 2923 86
NM_058176.2 NP_478056.1 21 - 22 2835 86
NM_178423.2 NP_848510.1 22 - 23 2980 86
NM_178425.3 NP_848512.1 21 - 22 2844 86
XM_011515629.2 XP_011513931.1 22 - 23 2938 86
XM_011515631.2 XP_011513933.1 23 - 24 3252 86
XM_011515634.2 XP_011513936.1 21 - 22 2806 86
XM_011515635.2 XP_011513937.1 23 - 24 3804 86
XM_011515636.2 XP_011513938.1 22 - 23 3525 86
XM_011515637.2 XP_011513939.1 22 - 23 2949 86
XM_011515638.2 XP_011513940.1 22 - 23 3519 86
XM_011515639.2 XP_011513941.1 22 - 23 3716 86
XM_011515641.2 XP_011513943.1 23 - 24 3801 86
XM_011515642.2 XP_011513944.1 21 - 22 4530 86
XM_011515644.2 XP_011513946.1 23 - 24 3776 86
XM_011515645.2 XP_011513947.1 21 - 22 4515 86
XM_011515646.2 XP_011513948.1 21 - 22 4506 86
XM_011515648.2 XP_011513950.1 22 - 23 3534 86
XM_011515649.2 XP_011513951.1 22 - 23 3738 86
XM_017012826.1 XP_016868315.1 22 - 23 3510 86
XM_017012827.1 XP_016868316.1 23 - 24 3593 86
XM_017012828.1 XP_016868317.1 21 - 22 3586 86
GenBank mRNA AJ459808.1 - 22 - 23 2944 86
AK095820.1 - 6 - 7 775 86
AY032737.1 - 21 - 22 2794 86
AY197371.1 - 21 - 22 2803 86
BC111735.1 - 20 - 21 2670 86

View Full Product Details