The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000823.3 | NP_000814.2 | 5 - 6 | 512 | 68 |
XM_011515263.1 | XP_011513565.1 | 2 - 3 | 530 | 68 | |
GenBank mRNA | AF282259.1 | - | 2 - 3 | 530 | 68 |
AF282260.1 | - | 2 - 3 | 530 | 68 | |
AF282261.1 | - | 3 - 4 | 354 | 68 | |
AY557192.1 | - | 5 - 6 | 464 | 68 | |
L01406.1 | - | 5 - 6 | 515 | 68 | |
L09237.1 | - | 5 - 6 | 512 | 68 |