The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015279.1 | NP_056094.1 | 1 - 2 | 462 | 100 |
XM_011538078.2 | XP_011536380.1 | 1 - 2 | 404 | 100 | |
XM_017019083.1 | XP_016874572.1 | 1 - 2 | 404 | 100 | |
GenBank mRNA | AB449914.1 | - | 1 - 2 | 154 | 100 |
AK055404.1 | - | 1 - 2 | 404 | 100 | |
AK299508.1 | - | 1 - 2 | 367 | 100 | |
DC377016.1 | - | 1 - 2 | 463 | 100 |