Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D30
Entrez Gene ID
23329
Gene Name
TBC1 domain family member 30
Gene Aliases
-
Chromosome Location
Chr.12: 64759489 - 64881022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015279.1 NP_056094.1 3 - 4 586 96
XM_011538078.2 XP_011536380.1 3 - 4 528 96
XM_017019083.1 XP_016874572.1 3 - 4 528 96
XM_017019084.1 XP_016874573.1 4 - 5 187 96
XM_017019085.1 XP_016874574.1 5 - 6 467 96
XM_017019086.1 XP_016874575.1 5 - 6 472 96
XM_017019087.1 XP_016874576.1 3 - 4 338 96
GenBank mRNA AB023201.1 - 4 - 5 181 96
AB449914.1 - 3 - 4 278 96
AK055404.1 - 3 - 4 528 96

View Full Product Details