Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D30
Entrez Gene ID
23329
Gene Name
TBC1 domain family member 30
Gene Aliases
-
Chromosome Location
Chr.12: 64759489 - 64881022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015279.1 NP_056094.1 6 - 7 1067 83
XM_011538078.2 XP_011536380.1 6 - 7 1009 83
XM_017019083.1 XP_016874572.1 6 - 7 1009 83
XM_017019084.1 XP_016874573.1 7 - 8 668 83
XM_017019085.1 XP_016874574.1 8 - 9 948 83
XM_017019086.1 XP_016874575.1 8 - 9 953 83
XM_017019087.1 XP_016874576.1 6 - 7 819 83
GenBank mRNA AB023201.1 - 7 - 8 662 83
AB449914.1 - 6 - 7 759 83
AK055404.1 - 6 - 7 1009 83

View Full Product Details