Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D30
Entrez Gene ID
23329
Gene Name
TBC1 domain family member 30
Gene Aliases
-
Chromosome Location
Chr.12: 64759489 - 64881022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017019084.1 XP_016874573.1 1 - 2 42 67
XM_017019085.1 XP_016874574.1 2 - 3 322 67
XM_017019086.1 XP_016874575.1 2 - 3 327 67
GenBank mRNA AB023201.1 - 1 - 2 36 67

View Full Product Details