The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152512.3 | NP_689725.2 | 1 - 2 | 99 | 104 |
XM_006724153.3 | XP_006724216.1 | 1 - 2 | 193 | 104 | |
XM_011529927.2 | XP_011528229.1 | 1 - 2 | 193 | 104 | |
XM_011529928.2 | XP_011528230.1 | 1 - 2 | 193 | 104 | |
XM_011529931.2 | XP_011528233.1 | 1 - 2 | 193 | 104 | |
GenBank mRNA | BC033895.2 | - | 1 - 2 | 99 | 104 |