The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152512.3 | NP_689725.2 | 5 - 6 | 1082 | 86 |
XM_006724149.3 | XP_006724212.1 | 5 - 6 | 1677 | 86 | |
XM_006724150.3 | XP_006724213.1 | 5 - 6 | 867 | 86 | |
XM_006724151.1 | XP_006724214.1 | 4 - 5 | 571 | 86 | |
XM_011529927.2 | XP_011528229.1 | 5 - 6 | 1176 | 86 | |
XM_011529928.2 | XP_011528230.1 | 4 - 5 | 933 | 86 | |
GenBank mRNA | AK093154.1 | - | 3 - 4 | 316 | 86 |
BC033895.2 | - | 5 - 6 | 1082 | 86 | |
CR456452.1 | - | 4 - 5 | 877 | 86 |