Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MBD6
Entrez Gene ID
114785
Gene Name
methyl-CpG binding domain protein 6
Gene Aliases
-
Chromosome Location
Chr.12: 57520710 - 57530148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052897.3 NP_443129.3 12 - 13 3165 94
XM_005268617.1 XP_005268674.1 12 - 13 3221 94
XM_005268618.1 XP_005268675.1 12 - 13 3317 94
XM_006719217.1 XP_006719280.1 12 - 13 3066 94
XM_006719218.1 XP_006719281.1 12 - 13 3054 94
XM_011537851.2 XP_011536153.1 12 - 13 4054 94
GenBank mRNA AB067474.1 - 9 - 10 2834 94
AK056399.1 - 12 - 13 3199 94
AK093078.1 - 7 - 8 2063 94
AL834230.1 - 6 - 7 1444 94
BC065530.1 - 12 - 13 3165 94
CR627478.1 - 5 - 6 744 94

View Full Product Details