Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 1 - 2 200 71
XM_005268725.3 XP_005268782.1 3 - 4 550 71
XM_011538037.1 XP_011536339.1 2 - 3 270 71
XM_017018998.1 XP_016874487.1 2 - 3 270 71
XM_017019001.1 XP_016874490.1 3 - 4 504 71
XM_017019002.1 XP_016874491.1 2 - 3 270 71
XM_017019005.1 XP_016874494.1 3 - 4 349 71
XM_017019006.1 XP_016874495.1 2 - 3 242 71
XM_017019007.1 XP_016874496.1 2 - 3 277 71
XM_017019010.1 XP_016874499.1 2 - 3 266 71
XM_017019013.1 XP_016874502.1 2 - 3 266 71
XM_017019014.1 XP_016874503.1 2 - 3 301 71
XM_017019016.1 XP_016874505.1 3 - 4 504 71
XM_017019017.1 XP_016874506.1 2 - 3 266 71
XM_017019018.1 XP_016874507.1 2 - 3 238 71
XM_017019021.1 XP_016874510.1 3 - 4 356 71
XM_017019022.1 XP_016874511.1 2 - 3 266 71
XM_017019023.1 XP_016874512.1 2 - 3 277 71
XM_017019024.1 XP_016874513.1 2 - 3 266 71
XM_017019025.1 XP_016874514.1 2 - 3 243 71
XM_017019027.1 XP_016874516.1 3 - 4 504 71
XM_017019028.1 XP_016874517.1 2 - 3 266 71
XM_017019029.1 XP_016874518.1 3 - 4 412 71
GenBank mRNA AB023219.1 - 3 - 4 557 71
AX775761.1 - 3 - 4 557 71
AX775763.1 - 3 - 4 557 71

View Full Product Details