Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 20 - 21 2536 70
XM_005268725.3 XP_005268782.1 22 - 23 2886 70
XM_011538033.1 XP_011536335.1 24 - 25 2828 70
XM_011538040.1 XP_011536342.1 23 - 24 2726 70
XM_017019000.1 XP_016874489.1 23 - 24 2732 70
XM_017019002.1 XP_016874491.1 22 - 23 2702 70
XM_017019003.1 XP_016874492.1 22 - 23 2678 70
XM_017019004.1 XP_016874493.1 22 - 23 2672 70
XM_017019005.1 XP_016874494.1 24 - 25 2883 70
XM_017019006.1 XP_016874495.1 23 - 24 2776 70
XM_017019007.1 XP_016874496.1 23 - 24 2811 70
XM_017019009.1 XP_016874498.1 22 - 23 2630 70
XM_017019011.1 XP_016874500.1 25 - 26 2927 70
XM_017019012.1 XP_016874501.1 21 - 22 2612 70
XM_017019013.1 XP_016874502.1 22 - 23 2746 70
XM_017019014.1 XP_016874503.1 22 - 23 2781 70
XM_017019016.1 XP_016874505.1 23 - 24 2942 70
XM_017019017.1 XP_016874506.1 22 - 23 2704 70
XM_017019018.1 XP_016874507.1 22 - 23 2670 70
XM_017019019.1 XP_016874508.1 23 - 24 2766 70
XM_017019020.1 XP_016874509.1 20 - 21 2516 70
XM_017019021.1 XP_016874510.1 22 - 23 2740 70
XM_017019022.1 XP_016874511.1 21 - 22 2650 70
XM_017019023.1 XP_016874512.1 21 - 22 2661 70
XM_017019024.1 XP_016874513.1 21 - 22 2650 70
XM_017019025.1 XP_016874514.1 21 - 22 2615 70
XM_017019026.1 XP_016874515.1 22 - 23 2655 70
XM_017019027.1 XP_016874516.1 21 - 22 2786 70
XM_017019028.1 XP_016874517.1 20 - 21 2548 70
XM_017019029.1 XP_016874518.1 20 - 21 2634 70
XM_017019030.1 XP_016874519.1 13 - 14 1915 70
XM_017019031.1 XP_016874520.1 16 - 17 2199 70
XM_017019032.1 XP_016874521.1 12 - 13 1767 70
XM_017019033.1 XP_016874522.1 14 - 15 1918 70
GenBank mRNA AB023219.1 - 22 - 23 2893 70
AK293394.1 - 12 - 13 1788 70
AX775761.1 - 22 - 23 2893 70
AX775763.1 - 22 - 23 2935 70
BC041857.1 - 12 - 13 2348 70
BC104995.1 - 10 - 11 1543 70
BC112226.1 - 10 - 11 1543 70
BC143605.1 - 11 - 12 1645 70

View Full Product Details