The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022916.4 | NP_075067.2 | 2 - 3 | 280 | 81 |
GenBank mRNA | AF439857.1 | - | 2 - 3 | 191 | 81 |
AK026840.1 | - | 2 - 3 | 191 | 81 | |
AK315545.1 | - | 2 - 3 | 218 | 81 | |
BC016617.1 | - | 2 - 3 | 188 | 81 | |
BI551252.1 | - | 2 - 3 | 233 | 81 | |
BP199163.1 | - | 2 - 3 | 280 | 81 |