The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022916.4 | NP_075067.2 | 4 - 5 | 594 | 123 |
GenBank mRNA | AF439857.1 | - | 4 - 5 | 505 | 123 |
AK315545.1 | - | 4 - 5 | 532 | 123 | |
BC016617.1 | - | 4 - 5 | 502 | 123 |