Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTLL9
Entrez Gene ID
164395
Gene Name
tubulin tyrosine ligase like 9
Gene Aliases
C20orf125
Chromosome Location
Chr.20: 31870702 - 31944959 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008409.2 NP_001008409.1 6 - 7 757 99
NR_134519.1 7 - 8 843 99
XM_005260309.2 XP_005260366.1 5 - 6 521 99
XM_006723724.2 XP_006723787.1 5 - 6 630 99
XM_011528617.1 XP_011526919.1 6 - 7 593 99
XM_011528618.1 XP_011526920.1 6 - 7 757 99
XM_011528619.1 XP_011526921.1 7 - 8 655 99
XM_011528620.2 XP_011526922.1 7 - 8 652 99
XM_011528621.1 XP_011526923.1 7 - 8 670 99
XM_011528622.1 XP_011526924.1 7 - 8 645 99
XM_011528623.2 XP_011526925.1 5 - 6 526 99
XM_011528625.1 XP_011526927.1 6 - 7 593 99
XM_011528626.1 XP_011526928.1 6 - 7 591 99
XM_011528627.2 XP_011526929.1 6 - 7 588 99
XM_011528628.1 XP_011526930.1 5 - 6 516 99
XM_011528629.1 XP_011526931.1 5 - 6 524 99
XM_011528630.1 XP_011526932.1 4 - 5 458 99
XM_011528632.1 XP_011526934.1 6 - 7 593 99
XM_011528633.1 XP_011526935.1 6 - 7 593 99
XM_011528634.1 XP_011526936.1 6 - 7 593 99
XM_011528635.1 XP_011526937.1 6 - 7 593 99
XM_011528637.2 XP_011526939.1 6 - 7 593 99
XM_011528638.1 XP_011526940.1 6 - 7 593 99
XM_011528639.1 XP_011526941.1 6 - 7 593 99
XM_011528640.1 XP_011526942.1 6 - 7 593 99
XM_011528641.1 XP_011526943.1 6 - 7 593 99
XM_011528643.2 XP_011526945.1 6 - 7 593 99
XM_011528644.1 XP_011526946.1 6 - 7 593 99
XM_017027695.1 XP_016883184.1 6 - 7 593 99
XM_017027696.1 XP_016883185.1 6 - 7 593 99
XM_017027697.1 XP_016883186.1 6 - 7 593 99
XM_017027698.1 XP_016883187.1 6 - 7 593 99
XM_017027699.1 XP_016883188.1 5 - 6 776 99
XM_017027700.1 XP_016883189.1 6 - 7 593 99
XM_017027701.1 XP_016883190.1 6 - 7 593 99
XM_017027702.1 XP_016883191.1 6 - 7 593 99
GenBank mRNA AK093491.1 - 6 - 7 758 99
AK302806.1 - 5 - 6 504 99
AK303963.1 - 5 - 6 395 99
BC068593.1 - 7 - 8 833 99
BC104025.1 - 5 - 6 630 99

View Full Product Details