The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135958.1 | NP_001129430.1 | 2 - 3 | 614 | 81 |
GenBank mRNA | AF421362.1 | - | 1 - 2 | 379 | 81 |