The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011535132.2 | XP_011533434.1 | 3 - 4 | 534 | 77 |
XM_017020640.1 | XP_016876129.1 | 3 - 4 | 549 | 77 |