The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016035.4 | NP_057119.2 | 3 - 4 | 650 | 80 |
GenBank mRNA | AF151850.1 | - | 3 - 4 | 621 | 80 |
AK025376.1 | - | 3 - 4 | 564 | 80 | |
AK301333.1 | - | 3 - 4 | 319 | 80 | |
AK313650.1 | - | 3 - 4 | 633 | 80 | |
BC011895.2 | - | 3 - 4 | 320 | 80 | |
CX787359.1 | - | 3 - 4 | 646 | 80 | |
EU216419.1 | - | 3 - 4 | 632 | 80 | |
EU216420.1 | - | 3 - 4 | 550 | 80 | |
EU216421.1 | - | 3 - 4 | 384 | 80 | |
EU216422.1 | - | 3 - 4 | 322 | 80 | |
EU216423.1 | - | 2 - 3 | 246 | 80 |