The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004443.3 | NP_004434.2 | 9 - 10 | 2236 | 78 |
GenBank mRNA | BC052968.1 | - | 9 - 10 | 2219 | 78 |
X75208.1 | - | 9 - 10 | 1826 | 78 |