The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021167.4 | NP_066990.3 | 3 - 4 | 712 | 111 |
NR_052016.1 | 3 - 4 | 712 | 111 | ||
GenBank mRNA | AB073650.1 | - | 1 - 2 | 806 | 111 |
AK026142.1 | - | 3 - 4 | 681 | 111 | |
AK098014.1 | - | 1 - 2 | 1038 | 111 | |
AK315788.1 | - | 3 - 4 | 678 | 111 | |
BC019350.1 | - | 3 - 4 | 683 | 111 | |
BC031091.1 | - | 3 - 4 | 586 | 111 |