Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPIP5K2
- Entrez Gene ID
-
23262
- Gene Name
- diphosphoinositol pentakisphosphate kinase 2
- Gene Aliases
-
CFAP160, HISPPD1, IP7K2, VIP2
- Chromosome Location
-
Chr.5: 103120254 - 103203523 on Build GRCh38
View Full Product Details