Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPIP5K2
Entrez Gene ID
23262
Gene Name
diphosphoinositol pentakisphosphate kinase 2
Gene Aliases
CFAP160, HISPPD1, IP7K2, VIP2
Chromosome Location
Chr.5: 103120254 - 103203523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276277.1 NP_001263206.1 26 - 27 3689 94
NM_001281471.1 NP_001268400.1 27 - 28 3734 94
NM_015216.3 NP_056031.2 25 - 26 3454 94
XM_005271935.1 XP_005271992.1 26 - 27 3688 94
XM_005271936.1 XP_005271993.1 25 - 26 3514 94
XM_005277534.1 XP_005277591.1 27 - 28 3733 94
XM_005277535.1 XP_005277592.1 27 - 28 3730 94
XM_005277536.1 XP_005277593.1 27 - 28 3733 94
XM_005277538.1 XP_005277595.1 26 - 27 3559 94
XM_017009269.1 XP_016864758.1 26 - 27 3685 94
XM_017009272.1 XP_016864761.1 25 - 26 3514 94
XM_017009273.1 XP_016864762.1 25 - 26 3511 94
GenBank mRNA AB007893.1 - 26 - 27 3679 94
AK294787.1 - 27 - 28 3598 94
BC024591.1 - 25 - 26 3207 94

View Full Product Details