The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276277.1 | NP_001263206.1 | 27 - 28 | 3813 | 110 |
NM_001281471.1 | NP_001268400.1 | 28 - 29 | 3858 | 110 | |
XM_005271936.1 | XP_005271993.1 | 26 - 27 | 3638 | 110 | |
XM_005277534.1 | XP_005277591.1 | 28 - 29 | 3857 | 110 | |
XM_005277535.1 | XP_005277592.1 | 28 - 29 | 3854 | 110 | |
XM_005277538.1 | XP_005277595.1 | 27 - 28 | 3683 | 110 | |
XM_017009269.1 | XP_016864758.1 | 27 - 28 | 3809 | 110 | |
GenBank mRNA | AB007893.1 | - | 27 - 28 | 3803 | 110 |
AK294787.1 | - | 28 - 29 | 3722 | 110 |