Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNK
Entrez Gene ID
8812
Gene Name
cyclin K
Gene Aliases
CPR4
Chromosome Location
Chr.14: 99481402 - 99511515 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005268154.4 XP_005268211.1 1 - 2 206 151

View Full Product Details