The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015278.3 | NP_056093.3 | 11 - 12 | 1759 | 106 |
XM_005266875.1 | XP_005266932.1 | 4 - 5 | 985 | 106 | |
XM_011535640.1 | XP_011533942.1 | 11 - 12 | 1202 | 106 | |
XM_017010598.1 | XP_016866087.1 | 11 - 12 | 2480 | 106 | |
XM_017010599.1 | XP_016866088.1 | 11 - 12 | 1803 | 106 | |
XM_017010600.1 | XP_016866089.1 | 11 - 12 | 1691 | 106 | |
XM_017010601.1 | XP_016866090.1 | 10 - 11 | 1905 | 106 | |
XM_017010602.1 | XP_016866091.1 | 9 - 10 | 1602 | 106 | |
XM_017010603.1 | XP_016866092.1 | 9 - 10 | 1541 | 106 | |
XM_017010604.1 | XP_016866093.1 | 8 - 9 | 1450 | 106 | |
XM_017010605.1 | XP_016866094.1 | 8 - 9 | 1416 | 106 | |
XM_017010606.1 | XP_016866095.1 | 4 - 5 | 1474 | 106 | |
XM_017010607.1 | XP_016866096.1 | 12 - 13 | 1382 | 106 | |
GenBank mRNA | AB018333.1 | - | 11 - 12 | 1502 | 106 |
AJ507735.1 | - | 11 - 12 | 1290 | 106 | |
AK024403.1 | - | 5 - 6 | 744 | 106 | |
AK025495.1 | - | 11 - 12 | 1759 | 106 | |
AK300403.1 | - | 4 - 5 | 669 | 106 | |
BC063279.1 | - | 11 - 12 | 1228 | 106 | |
BX537611.1 | - | 1 - 2 | 953 | 106 |