The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015278.3 | NP_056093.3 | 17 - 18 | 2686 | 72 |
XM_005266875.1 | XP_005266932.1 | 10 - 11 | 1912 | 72 | |
XM_011535640.1 | XP_011533942.1 | 17 - 18 | 2129 | 72 | |
XM_017010598.1 | XP_016866087.1 | 17 - 18 | 3407 | 72 | |
XM_017010599.1 | XP_016866088.1 | 17 - 18 | 2730 | 72 | |
XM_017010600.1 | XP_016866089.1 | 17 - 18 | 2618 | 72 | |
XM_017010601.1 | XP_016866090.1 | 16 - 17 | 2832 | 72 | |
XM_017010602.1 | XP_016866091.1 | 15 - 16 | 2529 | 72 | |
XM_017010603.1 | XP_016866092.1 | 15 - 16 | 2468 | 72 | |
XM_017010604.1 | XP_016866093.1 | 14 - 15 | 2377 | 72 | |
XM_017010605.1 | XP_016866094.1 | 14 - 15 | 2343 | 72 | |
XM_017010606.1 | XP_016866095.1 | 10 - 11 | 2401 | 72 | |
XM_017010607.1 | XP_016866096.1 | 18 - 19 | 2309 | 72 | |
GenBank mRNA | AB018333.1 | - | 17 - 18 | 2429 | 72 |
AJ507735.1 | - | 17 - 18 | 2217 | 72 | |
AK024403.1 | - | 11 - 12 | 1671 | 72 | |
AK025495.1 | - | 17 - 18 | 2686 | 72 | |
AK074249.1 | - | 7 - 8 | 971 | 72 | |
AK300403.1 | - | 10 - 11 | 1596 | 72 | |
BC063279.1 | - | 17 - 18 | 2155 | 72 | |
BX537611.1 | - | 6 - 7 | 1744 | 72 |