The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015278.3 | NP_056093.3 | 2 - 3 | 758 | 86 |
XM_011535640.1 | XP_011533942.1 | 2 - 3 | 201 | 86 | |
XM_017010598.1 | XP_016866087.1 | 2 - 3 | 990 | 86 | |
XM_017010599.1 | XP_016866088.1 | 2 - 3 | 313 | 86 | |
XM_017010600.1 | XP_016866089.1 | 2 - 3 | 201 | 86 | |
XM_017010607.1 | XP_016866096.1 | 2 - 3 | 316 | 86 | |
GenBank mRNA | AB018333.1 | - | 2 - 3 | 501 | 86 |
AJ507735.1 | - | 2 - 3 | 289 | 86 | |
AK025495.1 | - | 2 - 3 | 758 | 86 | |
BC063279.1 | - | 2 - 3 | 227 | 86 |