Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS12
Entrez Gene ID
57117
Gene Name
integrator complex subunit 12
Gene Aliases
INT12, PHF22, SBBI22
Chromosome Location
Chr.4: 105682627 - 105708725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142471.1 NP_001135943.1 1 - 2 84 73
GenBank mRNA AK314939.1 - 1 - 2 84 73

View Full Product Details