Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHC2
Entrez Gene ID
1912
Gene Name
polyhomeotic homolog 2
Gene Aliases
EDR2, HPH2, PH2
Chromosome Location
Chr.1: 33323623 - 33431017 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198040.2 NP_932157.1 3 - 4 465 101
XM_011540876.2 XP_011539178.1 4 - 5 934 101
XM_011540877.2 XP_011539179.1 4 - 5 934 101
XM_011540878.2 XP_011539180.1 4 - 5 934 101
XM_017000513.1 XP_016856002.1 4 - 5 895 101
XM_017000514.1 XP_016856003.1 4 - 5 507 101
XM_017000515.1 XP_016856004.1 4 - 5 600 101
XM_017000516.1 XP_016856005.1 4 - 5 895 101
XM_017000517.1 XP_016856006.1 4 - 5 507 101
XM_017000518.1 XP_016856007.1 4 - 5 600 101
XM_017000519.1 XP_016856008.1 4 - 5 904 101
XM_017000520.1 XP_016856009.1 4 - 5 507 101
XM_017000521.1 XP_016856010.1 4 - 5 926 101
XM_017000522.1 XP_016856011.1 4 - 5 934 101
GenBank mRNA AJ242730.1 - 4 - 5 504 101
AJ419231.1 - 3 - 4 465 101
AK292905.1 - 4 - 5 900 101
BC130630.1 - 3 - 4 465 101
BC144121.1 - 3 - 4 465 101

View Full Product Details