The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134439.1 | NP_001127911.1 | 1 - 2 | 152 | 123 |
GenBank mRNA | AB209903.1 | - | 1 - 2 | 33 | 123 |
AK092996.1 | - | 1 - 2 | 77 | 123 | |
AL832205.1 | - | 1 - 2 | 114 | 123 | |
BC038806.1 | - | 1 - 2 | 123 | 123 | |
DA272187.1 | - | 1 - 2 | 152 | 123 |