Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM1B
Entrez Gene ID
221656
Gene Name
lysine demethylase 1B
Gene Aliases
AOF1, C6orf193, LSD2, bA204B7.3, dJ298J15.2
Chromosome Location
Chr.6: 18155388 - 18223854 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005248925.1 XP_005248982.1 13 - 14 1598 125
XM_005248926.1 XP_005248983.1 13 - 14 1598 125
XM_005248928.1 XP_005248985.1 10 - 11 1163 125
XM_011514385.1 XP_011512687.1 11 - 12 1202 125
XM_011514386.1 XP_011512688.1 11 - 12 1202 125
XM_017010440.1 XP_016865929.1 14 - 15 1625 125
XM_017010441.1 XP_016865930.1 14 - 15 1625 125
XM_017010442.1 XP_016865931.1 13 - 14 1471 125
XM_017010443.1 XP_016865932.1 13 - 14 1418 125
XM_017010445.1 XP_016865934.1 12 - 13 1229 125

View Full Product Details