Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM1B
Entrez Gene ID
221656
Gene Name
lysine demethylase 1B
Gene Aliases
AOF1, C6orf193, LSD2, bA204B7.3, dJ298J15.2
Chromosome Location
Chr.6: 18155388 - 18223854 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153042.3 NP_694587.3 16 - 17 1778 154
XM_005248925.1 XP_005248982.1 20 - 21 2477 154
XM_005248926.1 XP_005248983.1 20 - 21 2474 154
XM_005248928.1 XP_005248985.1 17 - 18 2042 154
XM_011514385.1 XP_011512687.1 18 - 19 2081 154
XM_011514386.1 XP_011512688.1 18 - 19 2078 154
XM_017010440.1 XP_016865929.1 21 - 22 2504 154
XM_017010441.1 XP_016865930.1 21 - 22 2501 154
XM_017010442.1 XP_016865931.1 20 - 21 2350 154
XM_017010443.1 XP_016865932.1 20 - 21 2297 154
XM_017010445.1 XP_016865934.1 19 - 20 2108 154
GenBank mRNA AK091217.1 - 3 - 4 450 154
AK125318.1 - 16 - 17 1714 154
BC113093.1 - 3 - 4 209 154
CR627410.1 - 3 - 4 1361 154

View Full Product Details