Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM1B
Entrez Gene ID
221656
Gene Name
lysine demethylase 1B
Gene Aliases
AOF1, C6orf193, LSD2, bA204B7.3, dJ298J15.2
Chromosome Location
Chr.6: 18155388 - 18223854 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153042.3 NP_694587.3 2 - 3 229 71
XM_005248925.1 XP_005248982.1 2 - 3 229 71
XM_005248926.1 XP_005248983.1 2 - 3 229 71
XM_005248928.1 XP_005248985.1 2 - 3 229 71
XM_011514385.1 XP_011512687.1 2 - 3 229 71
XM_011514386.1 XP_011512688.1 2 - 3 229 71
XM_017010440.1 XP_016865929.1 2 - 3 229 71
XM_017010441.1 XP_016865930.1 2 - 3 229 71
XM_017010442.1 XP_016865931.1 2 - 3 165 71
XM_017010445.1 XP_016865934.1 2 - 3 229 71
XM_017010446.1 XP_016865935.1 2 - 3 229 71
GenBank mRNA AK125318.1 - 2 - 3 165 71
DA099422.1 - 2 - 3 230 71

View Full Product Details