The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166246.1 | NP_001159718.1 | 4 - 5 | 850 | 79 |
NM_021828.4 | NP_068600.4 | 4 - 5 | 850 | 79 | |
XM_006717937.2 | XP_006718000.1 | 4 - 5 | 910 | 79 | |
XM_011540029.1 | XP_011538331.1 | 4 - 5 | 850 | 79 | |
XM_011540030.1 | XP_011538332.1 | 3 - 4 | 675 | 79 | |
XM_011540031.2 | XP_011538333.1 | 3 - 4 | 459 | 79 | |
XM_011540033.2 | XP_011538335.1 | 3 - 4 | 483 | 79 | |
XM_017016495.1 | XP_016871984.1 | 4 - 5 | 850 | 79 | |
XM_017016496.1 | XP_016871985.1 | 4 - 5 | 850 | 79 | |
XM_017016497.1 | XP_016871986.1 | 3 - 4 | 443 | 79 | |
XM_017016498.1 | XP_016871987.1 | 3 - 4 | 744 | 79 | |
GenBank mRNA | AF282887.1 | - | 4 - 5 | 837 | 79 |
AJ299719.1 | - | 4 - 5 | 837 | 79 | |
AJ299720.1 | - | 4 - 5 | 777 | 79 | |
AK314759.1 | - | 4 - 5 | 850 | 79 | |
BC112356.1 | - | 4 - 5 | 819 | 79 |