Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr.5: 111092172 - 111130502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139281.2 NP_644810.1 13 - 14 1729 66
GenBank mRNA AF385437.1 - 13 - 14 1729 66
AK074202.1 - 7 - 8 779 66
AK095312.1 - 13 - 14 1587 66
AL832494.1 - 12 - 13 1531 66
BC133025.1 - 13 - 14 1689 66
BC136517.1 - 13 - 14 1689 66
CR749211.1 - 8 - 9 929 66

View Full Product Details