Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr.5: 111092172 - 111130502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139281.2 NP_644810.1 6 - 7 886 133
GenBank mRNA AF385437.1 - 6 - 7 886 133
AK095312.1 - 6 - 7 744 133
AL832494.1 - 5 - 6 692 133
BC133025.1 - 6 - 7 846 133
BC136517.1 - 6 - 7 846 133
CR749211.1 - 1 - 2 86 133

View Full Product Details