Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
Chromosome Location
Chr.1: 32862268 - 32872492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153756.2 NP_715637.2 5 - 6 700 91
GenBank mRNA AK092102.1 - 5 - 6 633 91

View Full Product Details