The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018206.5 | NP_060676.2 | 5 - 6 | 608 | 150 |
XM_005256045.3 | XP_005256102.1 | 4 - 5 | 493 | 150 | |
XM_011523227.2 | XP_011521529.1 | 5 - 6 | 1338 | 150 | |
GenBank mRNA | AF183418.1 | - | 4 - 5 | 517 | 150 |
AF186382.1 | - | 5 - 6 | 562 | 150 | |
AF191298.2 | - | 5 - 6 | 536 | 150 | |
AK001614.1 | - | 5 - 6 | 556 | 150 | |
AK023650.1 | - | 1 - 2 | 119 | 150 | |
AK223218.1 | - | 5 - 6 | 527 | 150 | |
AL136888.1 | - | 5 - 6 | 556 | 150 | |
BC002414.2 | - | 4 - 5 | 515 | 150 | |
BC010362.1 | - | 5 - 6 | 529 | 150 | |
BC093036.1 | - | 5 - 6 | 554 | 150 |